missing translation for 'onlineSavingsMsg'
Learn More
Learn More
Molecular Probes™ Monobromobimane (mBBr), 25 mg
Description
Monobromobimane is essentially nonfluorescent until conjugated, readily reacts with several low molecular weight thiols, including glutathione, N-acetylcysteine, mercaptopurine, peptides and plasma thiols. The glutathione conjugate of monobromobimane has absorption/emission maxima ∼394/490 nm.
Specifications
Specifications
| Quantity | 25 mg |
| Product Type | Monobromobimane |
| Content And Storage | Store in freezer (-5 to -30°C) and protect from light. |
| Shipping Condition | Room Temperature |
| Emission | 490 |
For Research Use Only. Not for use in diagnostic procedures.
Product Title
By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.
Spot an opportunity for improvement?