Learn More
Liebisch™ Monoblock Large Block Holder
Product Code. p-8014731
Click to view available options
For Use With (Equipment):
56 x 13.2 mm Tubes
Unit Size:
Each
Product Code. 11980108
Brand: Liebisch™ 15370000
This item is currently unavailable or has been discontinued.
View the product page for possible alternatives.
View alternative products
Specifications
For Use With (Equipment) 56 x 13.2 mm Tubes
Type Block Holder
Dimensions (L x W x H) 239 x 139 x 75 mm