Learn More
Mono-(7-carboxy-4-methylheptyl) Phthalate, TRC
Product Code. 30005741
Change view
Click to view available options
Quantity:
5 mg
50 mg
Unit Size:
50mg
5mg
2 product options available for selection
Product selection table with 2 available options. Use arrow keys to navigate and Enter or Space to select.
Product Code. Quantity unitSize
30005741 5 mg 5mg
30003580 50 mg 50mg
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 2 options available.
2 options
Product Code. 30005741 Supplier Toronto Research Chemicals Supplier No. TRCM5256055MG
Please to purchase this item. Need a web account? Register with us today!
High-purity organic molecules and analytical standards, strategically delivered worldwide to empower innovation and commercial success.
TRUSTED_SUSTAINABILITY
Chemical Identifiers
CAS 936022-02-5
Molecular Formula C17H22O6
Molecular Weight (g/mol) 322.35
Synonym 1,2-Benzenedicarboxylic Acid 1-(7-Carboxy-4-methylheptyl) Ester
SMILES O=C(C1=CC=CC=C1C(O)=O)OCCCC(C)CCCC(O)=O
Specifications
Quantity 5 mg
InChI Formula InChI=1S/C17H22O6/c1-12(6-4-10-15(18)19)7-5-11-23-17(22)14-9-3-2-8-13(14)16(20)21/h2-3,8-9,12H,4-7,10-11H2,1H3,(H,18,19)(H,20,21)
Formula Weight 322.1416
Percent Purity >95%
Chemical Name or Material Mono-(7-carboxy-4-methylheptyl) Phthalate
Grade HPLC