Learn More
Monensin sodium salt, MedChemExpress
Product Code. 16449006
Encompass_Preferred
Change view
Click to view available options
Quantity:
100 mg
Unit Size:
100mg
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Product Code. Quantity Unit Size
16449006 100 mg 100mg
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
Product Code. 16449006 Supplier Medchem Express Supplier No. HYN0150
 Restricted Item
Estimated Shipment: 04-09-2026
Add to basket
Add to basket
MedChemExpress Monensin sodium salt is an antibiotic secreted by the bacteria Streptomyces cinnamonensis. Monensin sodium salt is an ionophore that mediates Na+/H+ exchange. Monensin sodium salt causes a marked enlargement of the multivesicular bodies (MVBs) and regulates exosome secretion.
Monensin sodium salt is an antibiotic secreted by the bacteria Streptomyces cinnamonensis. Monensin sodium salt is an ionophore that mediates Na+/H+ exchange. Monensin sodium salt causes a marked enlargement of the multivesicular bodies (MVBs) and regulates exosome secretion.
Specifications
Chemical Name or Material Monensin sodium salt
Physical Form Solid
CAS 22373-78-0
Color White
Quantity 100 mg
Purity Grade Notes Research
Molecular Formula C36H61NaO11
For Use With (Application) COVID-19-immunoregulation
Synonym Monensin A sodium salt
Solubility Information Ethanol : 20 mg/mL (28.87 mM; Need ultrasonic) ∣DMSO : 5.4 mg/mL (7.79 mM; Need ultrasonic)
SMILES OC[C@@]1(O)[C@H](C)C[C@H](C)[C@]([C@@]2(O[C@]([C@@]3(CC[C@@]([C@]4(O[C@@]5(C[C@H](O)[C@@H](C)[C@@]([H])([C@H]([C@@H](OC)[C@H](C)C(O[Na])=O)C)O5)CC4)C)([H])O3)CC)([H])[C@@H](C)C2)[H])([H])O1
Molecular Weight (g/mol) 692.85
Formula Weight 692.85
Percent Purity 98.0%
Grade Research