missing translation for 'onlineSavingsMsg'
Learn More

Monensin sodium salt, 90% min., Thermo Scientific Chemicals

Product Code. p-7042201
Change view
Click to view available options
Quantity:
5 g
Unit Size:
5g
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Product Code. Quantity Unit Size
15494219 5 g 5g
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
This item is not returnable. View return policy
Product Code. 15494219 Supplier Thermo Scientific Alfa Aesar Supplier No. J61669.06

This item is currently unavailable or has been discontinued.
View the product page for possible alternatives.
View alternative products

This item is not returnable. View return policy

Monensin sodium salt is used in potentiometric and spectroscopic studies of alkali metal ion complexes. Monensin (Na) is a polyether small molecule ionophore, capable of forming stable complexes to monovalent cations with specific affinity for Na+. Monensin presents a highly lipophilic scaffold around the bound ion, allowing movement of the complex through the lipid bilayer and consequent transport of the ion out of the cell. Excessive flux of ions out of the cell produces a cytotoxic effect and generates the antibiotic properties of Monensin.

This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

Applications
Monensin sodium salt is used in potentiometric and spectroscopic studies of alkali metal ion complexes. Monensin (Na) is a polyether small molecule ionophore, capable of forming stable complexes to monovalent cations with specific affinity for Na+. Monensin presents a highly lipophilic scaffold around the bound ion, allowing movement of the complex through the lipid bilayer and consequent transport of the ion out of the cell. Excessive flux of ions out of the cell produces a cytotoxic effect and generates the antibiotic properties of Monensin.

Solubility
Soluble in methanol at 50mg/ml

Notes
Research Use only: Not intended for animal or human diagnostic or therapeutic use.
TRUSTED_SUSTAINABILITY

Chemical Identifiers

CAS 22373-78-0
Molecular Formula C36H61NaO11
Molecular Weight (g/mol) 692.863
MDL Number MFCD00077826
InChI Key XOIQMTLWECTKJL-JXDONITHSA-M
Synonym monensin, sodium salt, high purity, monensin sodium salt, monensin, monosodium salt, 4.5 dec-7-yl 3s,4r-3-methoxy-2-me thylpentanoic acid, sodium salt, 4-2-5-3r-5-3s,5r,6r-6-hydroxy-6-hydroxymethyl-3,5-dimethyl 2h-3,4,5, 6-tetrahydropyran-2-yl-3-methyloxolan-2-yl 5s-5-ethyloxolan-2-yl 5s,8s,2r ,9r-9-hydroxy-2,8-dimethyl-1,6-dioxaspiro, 4-2-2-ethyl-5'-6-hydroxy-6-hydroxymethyl-3,5-dimethyltetrahydro-2h-pyran-2-yl-3'-methyloctahydro-2,2'-bifuran-5-yl-9-hydroxy-2,8-dimethyl-1,6-dioxaspiro 4.5 decane-7-yl-3-methoxy-2-methylpentanoic acid sodium salt, potassium 4-2-5-ethyl-5-5-6-hydroxy-6-hydroxymethyl-3,5-dimethyloxan-2-yl-3-methyloxolan-2-yl oxolan-2-yl-9-hydroxy-2,8-dimethyl-1,6-dioxaspiro 4.5 decan-7-yl-3-methoxy-2-methylpentanoate
PubChem CID 133612076
IUPAC Name sodium;(3S,4R)-4-[(2R,5S,7R,8S,9R)-2-[(2S,5R)-5-ethyl-5-[(2S,3R,5S)-5-[(2R,3R,5S,6S)-6-hydroxy-6-(hydroxymethyl)-3,5-dimethyloxan-2-yl]-3-methyloxolan-2-yl]oxolan-2-yl]-7-hydroxy-2,8-dimethyl-1,10-dioxaspiro[4.5]decan-9-yl]-3-methoxy-2-methylpentanoate
SMILES CCC1(CCC(O1)C2(CCC3(O2)CC(C(C(O3)C(C)C(C(C)C(=O)[O-])OC)C)O)C)C4C(CC(O4)C5C(CC(C(O5)(CO)O)C)C)C.[Na+]

Specifications

Melting Point 267°C to 269°C
Color White
Quantity 5 g
UN Number UN3462
Beilstein 4122200
Merck Index 14,6246
Solubility Information Soluble in methanol at 50mg/ml
Formula Weight 692.86
Percent Purity 90 to 95.5%
Physical Form Powder
Chemical Name or Material Monensin sodium salt
Show More Show Less

RUO – Research Use Only

Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.