Learn More
Monensin Methyl Ester, TRC
Specifications
Specifications
| Chemical Name or Material | Monensin Methyl Ester |
| CAS | 28636-21-7 |
| Molecular Formula | C37 H64 O11 |
| Quantity | 50 mg |
| Synonym | 2-[5-Ethyltetrahydro-5-[tetrahydro-3-methyl-5-[tetrahydro-6-hydroxy-6-(hydroxymethyl)-3,5-dimethyl-2H-pyran-2-yl]-2-furyl]-2-furyl]-9-hydroxy-β-methoxy-α,γ,2,8-tetramethyl-1,6-dioxaspiro[4.5]decane-7-butyric Acid Methyl Ester, Monensin 1,6-Dioxaspiro[4.5]decane Deriv., Methyl Monensin, Monensin A Methyl Ester |
| InChI Formula | InChI=1S/C37H64O11/c1-11-35(32-21(3)17-27(44-32)29-20(2)16-22(4)37(41,19-38)47-29)13-12-28(45-35)34(8)14-15-36(48-34)18-26(39)23(5)31(46-36)24(6)30(42-9)25(7)33(40)43-10/h20-32,38-39,41H,11-19H2,1-10H3/t20-,21-,22+,23+,24-,25-,26-,27+,28+,29-,30+,31-,32+,34-,35-,36+,37-/m0/s1 |
| SMILES | CC[C@]1(CC[C@@H](O1)[C@]2(C)CC[C@]3(C[C@H](O)[C@@H](C)[C@H](O3)[C@@H](C)[C@@H](OC)[C@H](C)C(=O)OC)O2)[C@@H]4O[C@H](C[C@@H]4C)[C@H]5O[C@@](O)(CO)[C@H](C)C[C@@H]5C |
| IUPAC Name | methyl (2S,3R,4S)-4-[(2S,5R,7S,8R,9S)-2-[(2R,5S)-5-ethyl-5-[(2R,3S,5R)-5-[(2S,3S,5R,6R)-6-hydroxy-6-(hydroxymethyl)-3,5-dimethyloxan-2-yl]-3-methyloxolan-2-yl]oxolan-2-yl]-7-hydroxy-2,8-dimethyl-1,10-dioxaspiro[4.5]decan-9-yl]-3-methoxy-2-methylpentanoate |
| Molecular Weight (g/mol) | 684.898 |
| Formula Weight | 684.445 |