missing translation for 'onlineSavingsMsg'
Learn More

Hach™ Molybdovanadate Reagent

Product Code. 15558361
Click to view available options
Quantity:
100 mL
Unit Size:
Each
This item is not returnable. View return policy

Product Code. 15558361

Brand: Hach™ 2076032

Please to purchase this item. Need a web account? Register with us today!

This item is not returnable. View return policy

For phosphate (phosphorus) determination by the Molybdovanadate Method. In 100mL Marked Dropping Bottle.

Specifications

Color Yellow
Format Solution
For Use With (Equipment) All except DR850, DR820 and PC II
Packaging Dropper Bottle
Quantity 100 mL
Storage Requirements 10°C to 25°C
Test Method Molybdovanadate
Type Molybdovanadate Reagent
compliance-icons compliance-icons
Product Identifier
  • Molybdovanadate Reagent Solution
Signal Word
  • Danger
Hazard Category
  • Acute toxicity Category 4
  • Serious eye damage/eye irritation Category 1
  • Substances and mixtures corrosive to metals Category 1
  • Skin corrosion/irritation Category 1 A
Hazard Statement
  • H290-May be corrosive to metals.
  • H314-Causes severe skin burns and eye damage.
  • H318-Causes serious eye damage.
  • H332-Harmful if inhaled.
Precautionary Statement
  • P260-Do not breathe dust/fume/gas/mist/vapours/spray.
  • P280-Wear protective gloves/protective clothing/eye protection/face protection.
  • P301+P330+P331-IF SWALLOWED: rinse mouth. Do NOT induce vomiting.
  • P303+P361+P353-IF ON SKIN (or hair): Take off immediately all contaminated clothing. Rinse skin with water/ shower.
  • P304+P340-IF INHALED: Remove person to fresh air and keep comfortable for breathing.
  • P305+P351+P338-IF IN EYES: Rinse cautiously with water for several minutes. Remove contact lenses, if present and easy to do. Continue rinsing.
  • P310-Immediately call a POISON CENTER/doctor/.
Supplemental information
  • MIXTURE LIST-Contains : Sulfuric acid, Dipotassium peroxodisulphate
Product Content Correction

Your input is important to us. Please complete this form to provide feedback related to the content on this product.

Product Title

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.

Thank You! Your feedback has been submitted. Fisher Scientific is always working to improve our content for you. We appreciate your feedback.