missing translation for 'onlineSavingsMsg'
Learn More

Molibresib, MedChemExpress

Product Code. p-200101622
Encompass_Preferred
Click to view available options
Quantity:
5 mg
10 mg
50 mg
100 mg
200 mg
Unit Size:
100mg
10mg
200mg
50mg
5mg
This item is not returnable. View return policy

Product Code. 16476076

Brand: Medchem Express HY13032

Please to purchase this item. Need a web account? Register with us today!

This item is not returnable. View return policy

MedChemExpress Molibresib (I-BET762; GSK525762) is a BET bromodomain inhibitor with IC50 of 32.5-42.5 nM.

Molibresib (I-BET762

Specifications

Chemical Name or Material Molibresib
CAS 1260907-17-2
Color Light Yellow
Purity Grade Notes Research
Molecular Formula C22H22ClN5O2
Quantity 100 mg
Synonym I-BET762 GSK525762 GSK525762A
Solubility Information DMSO : 200 mg/mL (471.81 mM; Need ultrasonic)
SMILES ClC1=CC=C(C2=N[C@@H](CC(NCC)=O)C3=NN=C(C)N3C4=CC=C(OC)C=C24)C=C1
Molecular Weight (g/mol) 423.9
Formula Weight 423.9
Percent Purity 98.53%
Grade Research
Physical Form Solid
For Use With (Application) Cancer-programmed cell death
Show More Show Less

Research purposes only

Product Content Correction

The Fisher Scientific Encompass Program offers items which are not part of our distribution portfolio. These products typically do not have pictures or detailed descriptions. However, we are committed to improving your shopping experience. Please use the form below to provide feedback related to the content on this product.

Product Title

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.

Thank You! Your feedback has been submitted. Fisher Scientific is always working to improve our content for you. We appreciate your feedback.