Learn More
Molibresib, MedChemExpress
Product Code. 16476076
Encompass_Preferred
Click to view available options
Quantity:
5 mg
10 mg
50 mg
100 mg
200 mg
Unit Size:
100mg
10mg
200mg
50mg
5mg
Product Code. 16476076
Brand: Medchem Express HY13032
Please to purchase this item. Need a web account? Register with us today!
MedChemExpress Molibresib (I-BET762; GSK525762) is a BET bromodomain inhibitor with IC50 of 32.5-42.5 nM.
Molibresib (I-BET762
Specifications
Chemical Name or Material Molibresib
CAS 1260907-17-2
Color Light Yellow
Purity Grade Notes Research
Molecular Formula C22H22ClN5O2
Quantity 100 mg
Synonym I-BET762 GSK525762 GSK525762A
Solubility Information DMSO : 200 mg/mL (471.81 mM; Need ultrasonic)
SMILES ClC1=CC=C(C2=N[C@@H](CC(NCC)=O)C3=NN=C(C)N3C4=CC=C(OC)C=C24)C=C1
Molecular Weight (g/mol) 423.9
Formula Weight 423.9
Percent Purity 98.53%
Grade Research
Physical Form Solid
For Use With (Application) Cancer-programmed cell death