Learn More
Invitrogen™ Isolectin GS-IB4 From Griffonia simplicifolia, biotin-XX Conjugate
Description
Isolectin GS-IB4 is a 114,000-dalton glycoprotein that is part of a family of five tetrameric type I isolectins (IA4, IA3B, IA2B2, IAB3, and IB4). The A subunit prefers N-acetyl-D-galactosamine end groups while the B subunit is selective for terminal a-D-galactosyl residues. The biotin-XX isolectin GS-IB4 conjugate can be used in fluorescence microscopy or flow cytometry in conjunction with fluorescently labeled streptavidin.
Cell Analysis, Cellular Imaging, Flow Cytometry, Flow Cytometry Antibodies & Secondary Detection, Immunofluorescence (IF), Immunofluorescence Staining & Detection
Order Info
Shipping Condition: Room Temperature
Specifications
Specifications
| Quantity | 500 μg |
| Source | Africa Shrub legume |
| Conjugate | Biotin |
| Content And Storage | Store in freezer (-5 to -30°C). |
| Label Type | Biotin & Other Haptens |
| Shipping Condition | Room Temperature |
| Form | Lyophilized |
| Protein Subtype | Lectins |
For Research Use Only. Not for use in diagnostic procedures.
By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.