Learn More
Invitrogen™ DNP-BSA (Albumin from Bovine Serum (BSA), 2,4-Dinitrophenylated)
Product Code. 11538936
Change view
Click to view available options
Quantity:
5 mg
Unit Size:
5mg
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Product Code. Quantity Unit Size
11538936 5 mg 5mg
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
For Research Use Only. All usage must comply with product instructions.
Product Code. 11538936 Supplier Invitrogen™ Supplier No. A23018
 Restricted Item
In Stock
Add to basket
Add to basket
For Research Use Only. All usage must comply with product instructions.
Commonly used as protein tracer
  • Some applications of tracer include study of Fc receptor-mediated immune function and IgE- and IgG-mediated responses to crosslinking of DNP-specific antibodies
  • Because DNP-BSA has more defined molecular weight than polydisperse dextrans, it may be better suited for some applications where size of tracer protein must be known with accuracy
  • Has approximately 25 dinitrophenyl substituents per molecule of BSA
  • DNP-BSA can be detected with unlabeled or fluorescently labeled anti-DNP antibody
Cell Analysis, Cell Tracing and Tracking, Cellular Imaging, General Cell Tracing, Immunofluorescence (IF), Immunofluorescence Staining and Detection
Order Info
Shipping Condition: Room temperature
TRUSTED_SUSTAINABILITY
Specifications
Quantity 5 mg
Conjugate DNP (Dinitrophenyl)
Species Bovine
Content And Storage Store in freezer (-5 to -30°C) and protect from light.
Label Type Other Label(s) or Dye(s)
Shipping Condition Room Temperature
Form Lyophilized
Protein Subtype Albumin Proteins