Learn More
Invitrogen™ DiOC16(3) (3,3'-Dihexadecyloxacarbocyanine Perchlorate)
Description
The orange fluorescent, lipophilic carbocyanine DiOC16(3) is weakly fluorescent in water but highly fluorescent and quite photostable when incorporated into membranes. It has an extremely high extinction coefficient and short excited-state lifetimes (∽1 nanosecond) in lipid environments.
Cell Analysis, Cell Tracing & Tracking, Neuronal Tracing
Order Info
Shipping Condition: Room Temperature
Specifications
Specifications
| Color | Orange |
| Content And Storage | Store at room temperature and protect from light. |
| For Use With (Equipment) | Fluorescence Microscope |
| Quantity | 25 mg |
| Detection Method | Fluorescence |
| Shipping Condition | Room Temperature |
| Product Type | Liphophilic Tracer |
| SubCellular Localization | Cell Membranes, Lipids |
For Research Use Only. Not for use in diagnostic procedures.
By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.