Learn More
Invitrogen™ BAPTA, Tetracesium Salt, cell impermeant
Product Code. 10452632
Change view
Click to view available options
Quantity:
1 g
Unit Size:
1g
Product Code. Quantity unitSize
10452632 1 g 1g
1 options
For Research Use Only. All usage must comply with product instructions.
Product Code. 10452632
Brand: Invitrogen™ B1212
Please to purchase this item. Need a web account? Register with us today!
For Research Use Only. All usage must comply with product instructions.
A cell-impermeant chelator, which is a highly selective for Ca2+ over Mg2+
  • Can be used to control the level of extracellular Ca2+
  • BAPTA is more selective for Ca2+ than EDTA and EGTA, and its metal binding is also less sensitive to pH
Calcium Detection, Cell Analysis, Cell Viability, Proliferation and Function, Ionic Homeostasis and Signaling
Order Info
Shipping Condition: Room temperature
TRUSTED_SUSTAINABILITY
Specifications
Content And Storage Store at room temperature.
Product Type Cell-Impermeant Chelator
Dye Type Chelator (Non-Fluorescent)
Quantity 1 g
Shipping Condition Room Temperature