missing translation for 'onlineSavingsMsg'
Learn More

Mofegiline hydrochloride, MedChemExpress

Product Code. 16417146
Encompass_Preferred
Click to view available options
Quantity:
5 mg
10 mg
50 mg
100 mg
Unit Size:
100mg
10mg
50mg
5mg
This item is not returnable. View return policy

Product Code. 16417146

Brand: Medchem Express HY16677A

Please to purchase this item. Need a web account? Register with us today!

This item is not returnable. View return policy

MedChemExpress Mofegiline hydrochloride (MDL72974A) is a potent and selective enzyme-activated irreversible inhibitor of MAO-B; shows marked selectivity for the B form (IC50 = 680 and 3.6 nM for MAO-A and MAO-B).

Mofegiline hydrochloride (MDL72974A) is a potent and selective enzyme-activated irreversible inhibitor of MAO-B

Specifications

Chemical Name or Material Mofegiline hydrochloride
CAS 120635-25-8
Color Off-White
Purity Grade Notes Research
Molecular Formula C11H14ClF2N
Quantity 100 mg
Synonym MDL72974A
Solubility Information DMSO : 110 mg/mL (470.71 mM; Need ultrasonic) ∣H2O : 25 mg/mL (106.98 mM; Need ultrasonic)
SMILES FC1=CC=C(CC/C(CN)=C(F)/[H])C=C1.[H]Cl
Molecular Weight (g/mol) 233.69
Formula Weight 233.69
Percent Purity 95.0%
Grade Research
Physical Form Solid
For Use With (Application) Neuroscience-Neuromodulation
Show More Show Less

Research purposes only

Product Content Correction

The Fisher Scientific Encompass Program offers items which are not part of our distribution portfolio. These products typically do not have pictures or detailed descriptions. However, we are committed to improving your shopping experience. Please use the form below to provide feedback related to the content on this product.

Product Title

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.

Thank You! Your feedback has been submitted. Fisher Scientific is always working to improve our content for you. We appreciate your feedback.