missing translation for 'onlineSavingsMsg'
Learn More
Learn More
Moesin Mouse anti-Human, Rat, Clone: MSN/491, Novus Biologicals™
Mouse Monoclonal Antibody
Brand: Novus Biologicals NBP2-32875-20UG
This item is not returnable.
View return policy
Description
Moesin Monoclonal specifically detects Moesin in Human, Rat samples. It is validated for Western Blot, Simple Western, Flow Cytometry, Immunohistochemistry, Immunocytochemistry/Immunofluorescence, Immunohistochemistry-Paraffin.
Specifications
| Moesin | |
| Monoclonal | |
| Unconjugated | |
| PBS with 0.05% BSA. with 0.05% Sodium Azide | |
| Membrane-organizing extension spike protein, moesin | |
| Mouse | |
| Protein A purified | |
| RUO | |
| Primary | |
| Recognizes 78kDa moesin protein. Moesin, a member of the talin-4.1 superfamily, is a linking protein of the sub-membranous actin cytoskeleton. It is expressed in variable amounts in cells of different phenotypes such as macrophages, lymphocytes, fibroblastic, endothelial, epithelial, and neuronal cell lines but not in blood cells. The ERM proteins, ezrin, radixin, and moesin are involved in a variety of cellular functions, such as cell adhesion, migration, and the organization of cell surface structures, and are highly homologous, both in protein sequence and in functional activity, with merlin/schwannomin, a neurofibromatosis-2-associated tumor-suppressor protein. Cell lines of epithelial and mesothelial origin contain both moesin and radixin whereas cells of endothelial and lymphoid origin express moesin. | |
| Store at 4C. | |
| IgG1 κ |
| Western Blot, Flow Cytometry, Immunocytochemistry, Immunofluorescence, Immunoprecipitation | |
| MSN/491 | |
| Western Blot 0.5-1ug/ml, Simple Western 1.0 ug/ml, Flow Cytometry 0.5-1ug/million cells, Immunocytochemistry/Immunofluorescence 0.5-1ug/ml, Immunoprecipitation 0.5-1ug/500ug protein lysate, Immunohistochemistry-Paraffin 0.5-1.0ug/ml, Immunohistochemistry-Frozen 0.5-1.0ug/ml, SDS-Page | |
| P26038 | |
| MSN | |
| Recombinant human Moesin protein | |
| 20 μg | |
| Cytoskeleton Markers, Stem Cell Markers | |
| 4478 | |
| Human, Rat | |
| Purified |
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title
RUO
Spot an opportunity for improvement?Share a Content Correction