Learn More
Epredia™ Modified Grocott' s Methenamine Silver (Periodic Acid) Stain Kit
Shop All Epredia Products
Click to view available options
Type:
Fast Green Stain Solution
Gold Chloride Solution (0.1%)
Methenamine/Borax
Periodic Acid Schiff
Silver Nitrate Solution (5%)
Silver Stain Kit, Periodic Acid
Sodium Thiosulfate Solution
Unit Size:
125mL
30mL
Each
Pack of 12
Description
- Fungi, basement membranes and opportunistic organisms, along with reticular fibers, are stained black
- Background is stained green
Specifications
Specifications
| Type | Periodic Acid Schiff |
| Quantity | 125mL |
Safety and Handling
Signal Word
- Warning
- Skin corrosion/irritation Category 2
- Serious eye damage/eye irritation Category 2
- H315-Causes skin irritation.
- H319-Causes serious eye irritation.
- P264-Wash face, hands and anyexposed skin thoroughly after handling.
- P280-Wear protective gloves/protective clothing/eye protection/face protection.
- P302+P352-IF ON SKIN: Wash with plenty of water/soap.
- P332+P313-If skin irritation occurs: Get medical advice/attention.
- P361 + P364-Take off immediately all contaminated clothing and wash it before reuse.
- P305+P351+P338-IF IN EYES: Rinse cautiously with water for several minutes. Remove contact lenses, if present and easy to do. Continue rinsing.
- P337+P313-If eye irritation persists: Get medical advice/attention.
- MIXTURE LIST-Contains : periodic acid (H5IO6)