Learn More
Epredia™ Modified Grocott' s Methenamine Silver (Periodic Acid) Stain Kit
Product Code. 12583647 Shop All Epredia Products
Click to view available options
Type:
Fast Green Stain Solution
Gold Chloride Solution (0.1%)
Methenamine/Borax
Periodic Acid Schiff
Silver Nitrate Solution (5%)
Silver Stain Kit, Periodic Acid
Sodium Thiosulfate Solution
Unit Size:
125mL
30mL
Each
Pack of 12
Product Code. 12583647
Brand: Epredia™ 88026
Please to purchase this item. Need a web account? Register with us today!
This item is currently unavailable or has been discontinued.
View the product page for possible alternatives.
View alternative products
Distinguish fungi, basement membranes and opportunistic organisms such as Pneumocystis Carinii in tissue sections with Epredia™ Modified Grocott's Methenamine Silver (Periodic Acid)
  • Fungi, basement membranes and opportunistic organisms, along with reticular fibers, are stained black
  • Background is stained green
TRUSTED_SUSTAINABILITY
Specifications
Type Gold Chloride Solution (0.1%)
Quantity 125mL