Learn More
Modaline sulfate, MedChemExpress
Product Code. 16468596
Encompass_Preferred
Change view
Click to view available options
Quantity:
100 mg
Unit Size:
100mg
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Product Code. Quantity unitSize
16468596 100 mg 100mg
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
Product Code. 16468596 Supplier Medchem Express Supplier No. HYB1083
Please to purchase this item. Need a web account? Register with us today!
MedChemExpress Modaline sulfate is a MAO inhibitor, used in the treatment of depression.
Modaline sulfate is a MAO inhibitor, used in the treatment of depression.
Specifications
Chemical Name or Material Modaline sulfate
CAS 2856-75-9
Color Yellow
Purity Grade Notes Research
Molecular Formula C10H17N3O4S
Quantity 100 mg
Solubility Information DMSO : ≥ 31 mg/mL (112.60 mM)
SMILES CC1=NC=CN=C1N2CCCCC2.O=S(O)(O)=O
Molecular Weight (g/mol) 275.32
Formula Weight 275.32
Grade Research
Physical Form Solid
For Use With (Application) Neuroscience-Neuromodulation