Learn More
Modafinil, TRC
Specifications
Specifications
| Chemical Name or Material | Modafinil |
| CAS | 68693-11-8 |
| Purity Grade Notes | HPLC |
| Molecular Formula | C15 H15 N O2 S |
| Quantity | 50 mg |
| Synonym | Modafinil, 2-[(RS)-(Diphenylmethyl)sulfinyl]acetamide, Acetamide, 2-[(diphenylmethyl)sulfinyl]-, 2-[(Diphenylmethyl)sulfinyl]acetamide, (±)-Modafinil, 2-(Benzhydrylsulfinyl)acetamide, 2-(Benzhydrylsulfinyl)acetamide, 2-(Diphenyl-methanesulfinyl)-acetamide, 2-Diphenylmethanesulfinylacetamide, CEP 1538, CRC 40476, CRL 40476, DEP 1538, Modaphonil, Modiodal, Provigil |
| InChI Formula | InChI=1S/C15H15NO2S/c16-14(17)11-19(18)15(12-7-3-1-4-8-12)13-9-5-2-6-10-13/h1-10,15H,11H2,(H2,16,17) |
| SMILES | NC(=O)CS(=O)C(c1ccccc1)c2ccccc2 |
| IUPAC Name | 2-benzhydrylsulfinylacetamide |
| Molecular Weight (g/mol) | 273.35 |
| Show More |