missing translation for 'onlineSavingsMsg'
Learn More
Learn More
MNK2 Antibody - a Isoform, Novus Biologicals™
Rabbit Polyclonal Antibody
Brand: Novus Biologicals NBP2-60785
This item is not returnable.
View return policy
Description
MNK2 Polyclonal specifically detects MNK2 in Human, Primate samples. It is validated for Western Blot.
Specifications
| MNK2 | |
| Polyclonal | |
| Western Blot 1:100-1:2000 | |
| EC 2.7.11, EC 2.7.11.1, G protein-coupled receptor kinase 7, GPRK7Putative map kinase interacting kinase, MAP kinase interacting serine/threonine kinase 2, MAP kinase signal-integrating kinase 2, Mnk2, MNK2MAP kinase-interacting serine/threonine-protein kinase 2 | |
| Rabbit | |
| 47 kDa | |
| 100 μL | |
| Primary | |
| Human, Primate | |
| IgG |
| Western Blot | |
| Unconjugated | |
| 10 mM HEPES (pH 7.5), 150 mM NaCl, 100 ug per ml BSA and 50% glycerol. with No Preservative | |
| MKNK2 | |
| Synthetic peptide from the C-terminal region, specific to the human Mnk2a isoform. Accession # Q9HBH9 | |
| Antigen Affinity-purified | |
| Protein Kinase | |
| 2872 | |
| Store at -20C. Avoid freeze-thaw cycles. |
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title
Spot an opportunity for improvement?Share a Content Correction