missing translation for 'onlineSavingsMsg'
Learn More
Learn More
Description
MNK1 Polyclonal antibody specifically detects MNK1 in Human,Mouse,Rat samples. It is validated for ELISA,Immunohistochemistry,Western Blot,Immunohistochemistry (Paraffin),Immunocytochemistry/ Immunofluorescence
Specifications
Specifications
| Antigen | MNK1 |
| Applications | ELISA, Immunohistochemistry, Western Blot, Immunohistochemistry (Paraffin), Immunocytochemistry/Immunofluorescence |
| Classification | Polyclonal |
| Conjugate | Alexa Fluor 532 |
| Formulation | 50mM Sodium Borate |
| Gene Alias | EC 2.7.11, EC 2.7.11.1, MAP kinase interacting kinase 1, MAP kinase interacting serine/threonine kinase 1, MAP kinase signal-integrating kinase 1, MAP kinase-interacting serine/threonine-protein kinase 1, Mnk1, MNK1MAP kinase-interacting serine/threonine kinase 1 |
| Host Species | Rabbit |
| Immunogen | Recombinant fusion protein containing a sequence corresponding to amino acids 374-465 of human MNK1 (NP_003675.2).,, Sequence:, VQGQAPEKGLPTPQVLQRNSSTMDLTLFAAEAIALNRQLSQHEENELAEEPEALADGLCSMKLSPPCKSRLARRRALAQAGRGEDRSPPTAL |
| Purification Method | Affinity purified |
| Quantity | 0.1 mL |
| Show More |
Product Title
By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.
Spot an opportunity for improvement?