missing translation for 'onlineSavingsMsg'
Learn More

Tocris Bioscience™ MNI 137
GREENER_CHOICE

Product Code. 30001472
Click to view available options
Quantity:
10 mg
50 mg
Unit Size:
10mg
50mg
This item is not returnable. View return policy

Product Code. 30001472

Brand: Tocris Bioscience™ 4388/50

Please to purchase this item. Need a web account? Register with us today!

This item is not returnable. View return policy

Selective negative allosteric modulator of group II mGlu receptors

Selective negative allosteric modulator of group II mGlu receptors (IC50 values are 8.3 and 12.6 nM for human and rat mGlu2 inhibition of glutamate-induced calcium mobilization). Displays no activity at mGlu1, mGlu4, mGlu5 or mGlu8 receptors in a calcium mobilization assay.
TRUSTED_SUSTAINABILITY

Chemical Identifiers

CAS 946619-21-2
Molecular Formula C15H9BrN4O

Specifications

Quantity 50 mg
Formula Weight 341.16
Purity >98%
Molecular Weight (g/mol) 341.168
Chemical Name or Material MNI 137
Product Content Correction

Your input is important to us. Please complete this form to provide feedback related to the content on this product.

Product Title

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.

Thank You! Your feedback has been submitted. Fisher Scientific is always working to improve our content for you. We appreciate your feedback.