missing translation for 'onlineSavingsMsg'
Learn More
Learn More
MMP3 Monoclonal Antibody (1B4), Invitrogen™
Mouse Monoclonal Antibody
Brand: Invitrogen MA112893
Additional Details : Weight : 0.01000kg
Description
A suggested positive control for this product is esophageal or gastric tissues.
MMP3 (stromelysin-1) belongs to the peptidase M10A subfamily. It is secreted in its pro-protein form, and is activated by extracellular proteinase cleavage. MMP3 is capable of binding to zinc and calcium ions, and has metallo-endopeptidase activity. The active enzyme degrades collagen, gelatin, fibronectin, and laminin. It is involved in normal cellular processes such as wound repair and tissue remodeling. Mutations in the gene can result in coronary heart disease 6.Specifications
MMP3 | |
Monoclonal | |
1 mg/mL | |
PBS with 0.08% sodium azide | |
P08254 | |
MMP3 | |
Synthetic peptide derived from the C-terminus of the human MMP3 | |
200 μg | |
Primary | |
Human | |
Antibody | |
IgG1 |
ELISA, Immunohistochemistry, Western Blot | |
1B4 | |
Unconjugated | |
MMP3 | |
CHDS6; EC 3.4.24.17; EMS-2; matrix metallo protease; matrix metallopeptidase 3; matrix metallopeptidase 3 (stromelysin 1, progelatinase); matrix metalloproteinase 3; matrix metalloproteinase 3 (stromelysin 1, progelatinase); matrix metalloproteinase 3 precursor; matrix metalloproteinase-3; metalloproteinase 3 receptor; MMP; MMP10; MMP3; MMP-3; MMP-3 protein; MMPs; progelatinase; proteoglycanase; SL-1; SLN1; SLN-1; STMY; STMY1; Str1; STR-1; stromelysin; stromelysin 1; Stromelysin1; stromelysin-1; Transin1; Transin-1 | |
Mouse | |
Protein A/G | |
RUO | |
4314 | |
-20° C, Avoid Freeze/Thaw Cycles | |
Liquid |