Learn More
Invitrogen™ MMP17 Polyclonal Antibody

Rabbit Polyclonal Antibody
Brand: Invitrogen™ PA5116980
Description
Positive Samples: SW480, RAW264.7, Mouse testis, Rat testis Immunogen sequence: IMRPYYQGPV GDPLRYGLPY EDKVRVWQLY GVRESVSPTA QPEEPPLLPE PPDNRSSAPP RKDVPHRCST HFDAVAQIRG EAFFFKGKYF WRLTRDRHLV SLQPAQMHRF WRGLPLHLDS VDAVYERTSD HKIVFFKGDR YWVFKDNNVE EGYPRPVSDF SLPPGGIDAA FSWAHNDRTY FFKDQLYWRY DDHTRHMDPG YPAQSPLWRG VPSTLDDAMR WSDGASYFFR GQEYWKVLDG ELEVAPGYPQ STARDWLVCG DSQADGSVAA GVDAAEGPRA PPGQHDQSRS EDGYEVCSCT S.
Proteins of the matrix metalloproteinase (MMP) family are involved in the breakdown of extracellular matrix in normal physiological processes, such as embryonic development, reproduction, and tissue remodeling, as well as in disease processes, such as arthritis and metastasis. Most MMP's are secreted as inactive proproteins which are activated when cleaved by extracellular proteinases. The protein encoded by this gene is considered a member of the membrane-type MMP (MT-MMP) subfamily. However, this protein is unique among the MT-MMP's in that it is a GPI-anchored protein rather than a transmembrane protein. The protein activates MMP-2 by cleavage.
Specifications
| MMP17 | |
| Polyclonal | |
| Unconjugated | |
| MMP17 | |
| matrix metallo protease; matrix metallopeptidase 17; matrix metallopeptidase 17 (membrane-inserted); matrix metalloproteinase 17; matrix metalloproteinase 17 (membrane-inserted); matrix metalloproteinase-17; membrane type-4 matrix metalloproteinase; membrane-type matrix metalloproteinase 4; Membrane-type-4 matrix metalloproteinase; MMP; Mmp17; MMP-17; MMPs; MT4MMP; MT4-MMP; MT-MMP 4; MTMMP4 | |
| Rabbit | |
| Affinity chromatography | |
| RUO | |
| 23948, 288626, 4326 | |
| -20°C, Avoid Freeze/Thaw Cycles | |
| Liquid |
| ELISA, Immunohistochemistry (Paraffin), Western Blot | |
| 2.2 mg/mL | |
| PBS with 50% glycerol and 0.01% thimerosal; pH 7.3 | |
| Q9R0S3, Q9ULZ9 | |
| MMP17 | |
| Recombinant fusion protein containing a sequence corresponding to amino acids 265-565 of human MMP17 (NP_0572394). | |
| 100 μL | |
| Primary | |
| Human, Mouse, Rat | |
| Antibody | |
| IgG |
Your input is important to us. Please complete this form to provide feedback related to the content on this product.