missing translation for 'onlineSavingsMsg'
Learn More

Invitrogen™ MMP16 Polyclonal Antibody
GREENER_CHOICE

Product Code. 17711494
Change view
Click to view available options
Quantity:
100 μL
Unit Size:
100µL
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Product Code. Quantity unitSize
17711494 100 μL 100µL
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
This item is not returnable. View return policy
Product Code. 17711494 Supplier Invitrogen™ Supplier No. PA5120090

Please to purchase this item. Need a web account? Register with us today!

This item is not returnable. View return policy

Rabbit Polyclonal Antibody

Positive test controls include: Mouse brain, Mouse liver. The target is usually found in the following locations: Cell membrane, Cell surface, Extracellular side, Secreted, Single-pass type I membrane protein, extracellular matrix, extracellular space. Immunogen sequence: YGPPDKIPPP TRPLPTVPPH RSIPPADPRK NDRPKPPRPP TGRPSYPGAK PNICDGNFNT LAILRREMFV FKDQWFWRVR NNRVMDGYPM QITYFWRGLP PSIDAVYENS DGNFVFFKGN KYWVFKDTTL QPGYPHDLIT LGSGIPPHGI DSAIWWEDVG KTYFFKGDRY WRYSEEMKTM DPGYPKPITV WKGIPESPQG AFVHKENGFT YFYKGKEYWK FNNQILKVEP GYPRSILKDF MGCDGPTDRV KEGHSPPDDV DIVIKLDNTA STVKAI.

Matrix metallopeptidase 16 during embryonic development, reproduction, and tissue remodeling. MMP-16 is also involved in the ECM degradation processes associated with arthritis and metastasis. In humans, MMP-16 is a part of a cluster of MMP genes at the 11q22.3 locus.
TRUSTED_SUSTAINABILITY

Specifications

Antigen MMP16
Applications ELISA, Immunohistochemistry (Paraffin), Western Blot, Immunocytochemistry
Classification Polyclonal
Concentration 1.79 mg/mL
Conjugate Unconjugated
Formulation PBS with 50% glycerol and 0.09% sodium azide; pH 7.3
Gene Mmp16
Gene Accession No. P51512, Q9WTR0
Gene Alias C8orf57; DKFZp761D112; matrix metallo protease; matrix metallopeptidase 16; matrix metallopeptidase 16 (membrane-inserted); matrix metalloproteinase 16; matrix metalloproteinase-16; Membrane type 3-MMP; membrane-type matrix metalloproteinase 3; Membrane-type-3 matrix metalloproteinase; MMP; MMP-1; Mmp16; MMP-16; MMPs; MMPX2; MMP-X2; Mt3mmp; MT3-MMP; MT-MMP 3; MT-MMP2; MTMMP3; MT-MMP3; Putative transmembrane protein C8orf57
Gene Symbols Mmp16
Host Species Rabbit
Immunogen Recombinant fusion protein containing a sequence corresponding to amino acids 290-565 of human MMP16 (NP_005932.2).
Purification Method Affinity chromatography
Quantity 100 μL
Regulatory Status RUO
Primary or Secondary Primary
Gene ID (Entrez) 17389, 4325
Target Species Human, Mouse
Content And Storage -20°C, Avoid Freeze/Thaw Cycles
Product Type Antibody
Form Liquid
Isotype IgG
Show More Show Less
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.