missing translation for 'onlineSavingsMsg'
Learn More
Learn More
MLLT4, Rabbit anti-Human, Polyclonal Antibody, Abnova™
Rabbit Polyclonal Antibody
Brand: Abnova PAB31146.100uL
This item is not returnable.
View return policy
Description
Sequence: DSGGTLRIYADSLKPNIPYKTILLSTTDPADFAVAEALEKYGLEKENPKDYCIARVMLPPGAQHSDEKGAKEIILDDDECPSpecifications
| MLLT4 | |
| Polyclonal | |
| Immunohistochemistry (Formalin/PFA-fixed paraffin-embedded sections) (1:200-1:500) The optimal working dilution should be determined by the end user. | |
| myeloid/lymphoid or mixed-lineage leukemia (trithorax homolog, Drosophila); translocated to, 4 | |
| MLLT4 | |
| Recombinant protein corresponding to human MLLT4. | |
| RUO | |
| 4301 | |
| Store at 4°C for short term storage. For long term storage store at -20°C. Aliquot to avoid repeated freezing and thawing. | |
| Liquid |
| Immunohistochemistry (Paraffin) | |
| Unconjugated | |
| In PBS, pH 7.2 (40% glycerol, 0.02% sodium azide). | |
| AF-6/AF6/AFADIN/FLJ34371/RP3-431P23.3 | |
| Rabbit | |
| 100 μL | |
| Primary | |
| Human | |
| Antibody | |
| IgG |
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title
Spot an opportunity for improvement?Share a Content Correction