Learn More
MLH1 Antibody (S02-2E8), Novus Biologicals™
Rabbit Monoclonal Antibody
Brand:  Novus Biologicals NBP3-19527-50ul
 View more versions of this product
Product Code. 18621194
  • 421.00 € / 50µL
Please to purchase this item. Need a web account? Register with us today!
This item is not returnable. View return policy
Description
Description
MLH1 Monoclonal antibody specifically detects MLH1 in Human, Mouse, Rat samples. It is validated for Western Blot, Immunocytochemistry/ Immunofluorescence
TRUSTED_SUSTAINABILITY
Specifications
Specifications
MLH1
Monoclonal
0.3 mg/mL
50mM Tris-Glycine(pH 7.4), 0.15M NaCl, 40% Glycerol, 0.05% BSA
Rabbit
Affinity purified
RUO
Primary
Human, Mouse, Rat
Purified
Western Blot, Immunofluorescence
S02-2E8
Unconjugated
COCA2FCC2, DNA mismatch repair protein Mlh1, hMLH1, HNPCC, HNPCC2MGC5172, mutL (E. coli) homolog 1 (colon cancer, nonpolyposis type 2), mutL homolog 1, colon cancer, nonpolyposis type 2 (E. coli), MutL protein homolog 1
A synthetic peptide of human MLH1 (Uniprot # P40692)
50 μg
Apoptosis, Breast Cancer, Cancer, Cell Cycle and Replication, Checkpoint signaling, DNA Double Strand Break Repair, DNA Repair, Mismatch Repair, Tumor Suppressors
4292
Store at 4°C short term. Store at -20°C long term. Avoid freeze-thaw cycles.