Learn More
ML367, MedChemExpress
Product Code. 16465776
Encompass_Preferred
Click to view available options
Quantity:
50 mg
100 mg
Unit Size:
100mg
50mg
Product Code. 16465776
Brand: Medchem Express HY122198
Please to purchase this item. Need a web account? Register with us today!
MedChemExpress ML367 is a potent inhibitor of ATPase family AAA domain-containing protein 5 (ATAD5) stabilization, acts as a probe molecule that has low micromolar inhibitory activity. ML367 blocks DNA repair pathways, suppresses general DNA damage responses including RPA32-phosphorylation and CHK1-phosphorylation in response to UV irradiation.
ML367 is a potent inhibitor of ATPase family AAA domain-containing protein 5 (ATAD5) stabilization, acts as a probe molecule that has low micromolar inhibitory activity. ML367 blocks DNA repair pathways, suppresses general DNA damage responses including RPA32-phosphorylation and CHK1-phosphorylation in response to UV irradiation.
Specifications
Chemical Name or Material ML367
CAS 381168-77-0
Color Light Yellow
Purity Grade Notes Research
Molecular Formula C19H12F2N4
Quantity 100 mg
Solubility Information DMSO : ≥ 125 mg/mL (373.89 mM)
SMILES FC1=CC(NC2=NC(C3=CC=NC=C3)=NC4=CC=CC=C42)=CC=C1F
Molecular Weight (g/mol) 334.32
Formula Weight 334.32
Percent Purity 99.84%
Grade Research
Physical Form Solid
For Use With (Application) Neuroscience-Neuromodulation