missing translation for 'onlineSavingsMsg'
Learn More

MKT-077, MedChemExpress

Product Code. p-200101834
Encompass_Preferred
Click to view available options
Quantity:
5 mg
10 mg
Unit Size:
10mg
5mg
This item is not returnable. View return policy

Product Code. 16468897

Brand: Medchem Express HY15096

Please to purchase this item. Need a web account? Register with us today!

This item is not returnable. View return policy

MedChemExpress MKT-077 is a rhodacyanine dye and also a heat shock protein 70 (Hsp70) inhibitor which exhibits significant antitumor activity.

MKT-077 is a rhodacyanine dye and also a heat shock protein 70 (Hsp70) inhibitor which exhibits significant antitumor activity.

Specifications

Chemical Name or Material MKT-077
CAS 147366-41-4
Purity Grade Notes Research
Molecular Formula C21H22ClN3OS2
Synonym FJ-776
Solubility Information DMSO : 56.67 mg/mL (131.18 mM; Need ultrasonic) ∣H2O : < 0.1 mg/mL (ultrasonic) (insoluble)
SMILES CN(C1=CC=CC=C1S/2)C2=C3C(N(CC)/C(S\3)=C/C4=[N+](CC)C=CC=C4)=O.[Cl-]
Molecular Weight (g/mol) 432.0
Formula Weight 432.0
Percent Purity 98.0%
Grade Research
Color Brown
Quantity 5 mg
Physical Form Solid
Show More Show Less

Research purposes only

Product Content Correction

The Fisher Scientific Encompass Program offers items which are not part of our distribution portfolio. These products typically do not have pictures or detailed descriptions. However, we are committed to improving your shopping experience. Please use the form below to provide feedback related to the content on this product.

Product Title

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.

Thank You! Your feedback has been submitted. Fisher Scientific is always working to improve our content for you. We appreciate your feedback.