missing translation for 'onlineSavingsMsg'
Learn More

MK-8245, MedChemExpress

Product Code. 16431707
Encompass_Preferred
Click to view available options
Quantity:
2 mg
5 mg
10 mg
50 mg
100 mg
Unit Size:
100mg
10mg
2mg
50mg
5mg
This item is not returnable. View return policy

Product Code. 16431707

Brand: Medchem Express HY13070

Please to purchase this item. Need a web account? Register with us today!

This item is not returnable. View return policy

MedChemExpress MK-8245 is a potent, liver-targeted stearoyl-CoA desaturase (SCD) inhibitor, with IC50s of 1 nM for human SCD1 and 3 nM for both rat SCD1 and mouse SCD1, with antidiabetic and antidyslipidemic efficacy.

MK-8245 is a potent, liver-targeted stearoyl-CoA desaturase (SCD) inhibitor, with IC50s of 1 nM for human SCD1 and 3 nM for both rat SCD1 and mouse SCD1, with antidiabetic and antidyslipidemic efficacy.

Specifications

Chemical Name or Material MK-8245
CAS 1030612-90-8
Color White
Purity Grade Notes Research
Molecular Formula C17H16BrFN6O4
Quantity 50 mg
Solubility Information DMSO : ≥ 100 mg/mL (214.02 mM)
SMILES O=C(CN1N=C(N=N1)C2=CC(N3CCC(CC3)OC4=CC(F)=CC=C4Br)=NO2)O
Molecular Weight (g/mol) 467.25
Formula Weight 467.25
Percent Purity 98.23%
Grade Research
Physical Form Solid
For Use With (Application) Metabolism-sugar/lipid metabolism
Show More Show Less

Research purposes only

Product Content Correction

The Fisher Scientific Encompass Program offers items which are not part of our distribution portfolio. These products typically do not have pictures or detailed descriptions. However, we are committed to improving your shopping experience. Please use the form below to provide feedback related to the content on this product.

Product Title

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.

Thank You! Your feedback has been submitted. Fisher Scientific is always working to improve our content for you. We appreciate your feedback.