missing translation for 'onlineSavingsMsg'
Learn More
Learn More
MK-8245, MedChemExpress
Click to view available options
Quantity:
2 mg
5 mg
10 mg
50 mg
100 mg
Unit Size:
100mg
10mg
2mg
50mg
5mg
Description
MK-8245 is a potent, liver-targeted stearoyl-CoA desaturase (SCD) inhibitor, with IC50s of 1 nM for human SCD1 and 3 nM for both rat SCD1 and mouse SCD1, with antidiabetic and antidyslipidemic efficacy.
Specifications
Specifications
| Chemical Name or Material | MK-8245 |
| CAS | 1030612-90-8 |
| Color | White |
| Purity Grade Notes | Research |
| Molecular Formula | C17H16BrFN6O4 |
| Quantity | 10 mg |
| Solubility Information | DMSO : ≥ 100 mg/mL (214.02 mM) |
| SMILES | O=C(CN1N=C(N=N1)C2=CC(N3CCC(CC3)OC4=CC(F)=CC=C4Br)=NO2)O |
| Molecular Weight (g/mol) | 467.25 |
| Formula Weight | 467.25 |
| Show More |
Research purposes only
Product Content Correction
The Fisher Scientific Encompass Program offers items which are not part of our distribution portfolio. These products typically do not have pictures or detailed descriptions. However, we are committed to improving your shopping experience. Please use the form below to provide feedback related to the content on this product.
Product Title
Ser du en mulighed for forbedring?Del en indholdskorrektion