missing translation for 'onlineSavingsMsg'
Learn More

MK-8033, MedChemExpress

Product Code. p-200096423
Encompass_Preferred
Click to view available options
Quantity:
5 mg
10 mg
50 mg
Unit Size:
10mg
50mg
5mg
This item is not returnable. View return policy

Product Code. 16495628

Brand: Medchem Express HY13299

Please to purchase this item. Need a web account? Register with us today!

This item is not returnable. View return policy

MedChemExpress MK-8033 is an orally active ATP competitive c-Met/Ron dual inhibitor (IC50s: 1 nM (c-Met),7 nM (Ron)), with preferential binding to the activated kinase conformation. MK-8033 can be used in the research of cancers, such as breast and bladder cancers, non-small cell lung cancers (NSCLCs).

MK-8033 is an orally active ATP competitive c-Met/Ron dual inhibitor (IC50s: 1 nM (c-Met),7 nM (Ron)), with preferential binding to the activated kinase conformation. MK-8033 can be used in the research of cancers, such as breast and bladder cancers, non-small cell lung cancers (NSCLCs).

Specifications

Chemical Name or Material MK-8033
CAS 1001917-37-8
Color Light Yellow
Purity Grade Notes Research
Molecular Formula C25H21N5O3S
Quantity 10 mg
Solubility Information DMSO : ≥ 46 mg/mL (97.55 mM)
SMILES O=S(NCC1=CC=CC=N1)(CC2=CC=C3C(C(C4=CC(C5=CN(N=C5)C)=CN=C4C=C3)=O)=C2)=O
Molecular Weight (g/mol) 471.53
Formula Weight 471.53
Grade Research
Physical Form Solid
For Use With (Application) Cancer-Kinase/protease
Show More Show Less

Research purposes only

Product Content Correction

The Fisher Scientific Encompass Program offers items which are not part of our distribution portfolio. These products typically do not have pictures or detailed descriptions. However, we are committed to improving your shopping experience. Please use the form below to provide feedback related to the content on this product.

Product Title

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.

Thank You! Your feedback has been submitted. Fisher Scientific is always working to improve our content for you. We appreciate your feedback.