Learn More
MK-0773, MedChemExpress
Click to view available options
Quantity:
5 mg
10 mg
50 mg
100 mg
Unit Size:
100mg
10mg
50mg
5mg
Description
MK-0773 is a selective androgen receptor modulators (SARMs) that binds to AR with an IC50 of 6.6 nM.
Specifications
Specifications
| Chemical Name or Material | MK-0773 |
| Physical Form | Solid |
| CAS | 606101-58-0 |
| Color | Light Yellow |
| Quantity | 10 mg |
| Purity Grade Notes | Research |
| Molecular Formula | C27H34FN5O2 |
| Synonym | PF-05314882 |
| Solubility Information | DMSO : 33.33 mg/mL (69.50 mM; Need ultrasonic) |
| SMILES | O=C([C@H]1CC[C@]2([H])[C@]1(C)CC[C@]3([H])[C@@]4(C)C=C(F)C(N(C)[C@]4([H])CC[C@]32[H])=O)NCC5=NC6=CC=CN=C6N5 |
| Show More |