missing translation for 'onlineSavingsMsg'
Learn More

MK-0752, MedChemExpress

Product Code. 16424328
Encompass_Preferred
Click to view available options
Quantity:
5 mg
10 mg
50 mg
100 mg
Unit Size:
100mg
10mg
50mg
5mg
This item is not returnable. View return policy

Product Code. 16424328

Brand: Medchem Express HY10974

Please to purchase this item. Need a web account? Register with us today!

This item is not returnable. View return policy

MedChemExpress MK-0752 is a potent, orally active and specific γ-secretase inhibitor, showing dose-dependent reduction of Aβ40 with an IC50 of 5 nM in human SH-SY5Y cells. MK-0752 crosses the blood-brain barrier. MK-0752 reduces newly generated CNS Aβ in vivo.

MK-0752 is a potent, orally active and specific γ-secretase inhibitor, showing dose-dependent reduction of Aβ40 with an IC50 of 5 nM in human SH-SY5Y cells. MK-0752 crosses the blood-brain barrier. MK-0752 reduces newly generated CNS Aβ in vivo.

Specifications

Chemical Name or Material MK-0752
CAS 471905-41-6
Color White
Purity Grade Notes Research
Molecular Formula C21H21ClF2O4S
Quantity 10 mg
Solubility Information DMSO : ≥ 100 mg/mL (225.78 mM) ∣Ethanol : 10 mg/mL (22.58 mM; Need ultrasonic)
SMILES O=S([C@@]1(C2=C(C=CC(F)=C2)F)CC[C@@H](CCC(O)=O)CC1)(C3=CC=C(Cl)C=C3)=O
Molecular Weight (g/mol) 442.9
Formula Weight 442.9
Percent Purity 95.0%
Grade Research
Physical Form Solid
For Use With (Application) Cancer-programmed cell death
Show More Show Less

Research purposes only

Product Content Correction

The Fisher Scientific Encompass Program offers items which are not part of our distribution portfolio. These products typically do not have pictures or detailed descriptions. However, we are committed to improving your shopping experience. Please use the form below to provide feedback related to the content on this product.

Product Title

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.

Thank You! Your feedback has been submitted. Fisher Scientific is always working to improve our content for you. We appreciate your feedback.