Learn More
Invitrogen™ Mitochondria Monoclonal Antibody (MTC02)

Mouse Monoclonal Antibody
Brand: Invitrogen™ MA512017
Description
This antibody recognizes a 60kDa non-glycosylated protein component of mitochondria found in normal and malignant human cells. Staining of formalin-fixed tissues requires boiling tissue sections in 10mM citrate buffer, pH 6.0 for 10-20 min followed by cooling at room temperature for 20 min. Recommended positive controls: HeLa cells and liver tissue.
Antibodies that recognize subcellular organelles or compartments of human cells are useful in identification of these organelles in cells, tissues, and biochemical preparations. Anti-mitochondria antibodies can be used to stain mitochondria in cell/tissue preparations and can be used as a marker in subcellular fractions.
Specifications
| Mitochondria | |
| Monoclonal | |
| 0.2 mg/mL | |
| PBS with 0.05% BSA and 0.05% sodium azide; pH 7.4 | |
| Semi-purified mitochondrial preparation. | |
| 500 μL | |
| Primary | |
| 4°C | |
| Liquid |
| Immunohistochemistry (Paraffin), Western Blot, Immunocytochemistry | |
| MTC02 | |
| Unconjugated | |
| Mouse | |
| Protein A/G | |
| RUO | |
| Human | |
| Antibody | |
| IgG1 κ |
Your input is important to us. Please complete this form to provide feedback related to the content on this product.