missing translation for 'onlineSavingsMsg'
Learn More

Tocris Bioscience™ MitoBrilliant Live 646

Product Code. p-200085197
Change view
Click to view available options
Quantity:
50 μg (Trial size)
Unit Size:
50µg
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Product Code. Quantity unitSize
30040308 50 μg (Trial size) 50µg
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
This item is not returnable. View return policy
Product Code. 30040308 Supplier Tocris Bioscience™ Supplier No. 7417/50U

Please to purchase this item. Need a web account? Register with us today!

This item is not returnable. View return policy

Red fluorescent mitochondrial stain for live cells, Δψm dependent

MitoBrilliant probes have been developed to overcome some common limitations encountered with standard mitochondrial trackers, offering clearer answers to scientific questions. They are next-generation fluorescent stains for the localization and tracking of mitochondria in both live and fixed cells. The range harnesses Janelia Fluor® dye technology, conferring some of the properties that make these widely used dyes, into mitochondrial stains.

The MitoBrilliant™ Live dyes accumulate in the mitochondria of live cells in a mitochondrial membrane potential (Δψm) dependent manner. Upon loss of the mitochondrial membrane potential, the dyes disperse, and so provide a dynamic assessment of the mitochondrial membrane potential in live cells. Two dyes are available: MitoBrilliant™ Live 646 (red emission) and MitoBrilliant™ Live 549 (yellow/orange emission).

A universal stain for mitochondria in both live and fixed cells is also available: MitoBrilliant™ 646 .

Please refer to the protocol for guidelines on product use, and download the MitoBrilliant Product Guide to view data for each of the dyes in different applications.

MitoBrilliant is a trademark of Bio-Techne Corp.

TRUSTED_SUSTAINABILITY

Specifications

Quantity 50 μg (Trial size)
Target Mitochondria Dyes and Probes
Purity 95%
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.