Learn More
Schmidlin™ Mistral Evolution Nitrogen Generator
Product Code. 17718879
Encompass_Preferred
Click to view available options
Flow Rate:
10 L/min.
25 L/min.
40 L/min.
Nitrogen Purity:
>97%
>99%
>99.5%
Outlet Pressure:
3 bar
8 bar
Unit Size:
Each
Product Code. 17718879
Brand: Schmidlin™ DBEVO10EU
Please to purchase this item. Need a web account? Register with us today!
This generator produces a continuous supply of high purity nitrogen.
  • Increases efficiency and return on investment
  • On-demand supply 24/7
  • Flow rate at 8 barg (116 psig)
  • LC/MS grade purity
  • Integrated, low noise, oil-free compressors
  • Proprietary carbon molecular sieve technology
  • Complete 'plug and play' LC/MS solution
Specifications
Depth (Metric) 83.5 cm
Flow Rate 10 L/min.
Height (Metric) 68 cm
Max. Ambient Operating Temperature 35°C
Outlet Pressure 3 bar
Width (English) 19 in.
Width (Metric) 48.5 cm
Connections Outlet: 6 mm OD Compression or 1/4 in.
Electrical Requirements 110/120 V at 60 Hz, 220/240 V at 50 Hz
Nitrogen Purity >99.5%
Wattage 1800 W