Learn More
Miridesap, MedChemExpress
Product Code. 16446347
Encompass_Preferred
Change view
Click to view available options
Quantity:
5 mg
Unit Size:
5mg
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Product Code. Quantity unitSize
16446347 5 mg 5mg
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
Product Code. 16446347 Supplier Medchem Express Supplier No. HY101861
Please to purchase this item. Need a web account? Register with us today!
MedChemExpress Miridesap is a ligand for serum amyloid P component (SAP) and intends to inhibit and dissociate SAP binding to amyloid fibrils and tangles.
Miridesap is a ligand for serum amyloid P component (SAP) and intends to inhibit and dissociate SAP binding to amyloid fibrils and tangles.
Specifications
Chemical Name or Material Miridesap
CAS 224624-80-0
Color White
Purity Grade Notes Research
Molecular Formula C16H24N2O6
Quantity 5 mg
Synonym CPHPC Ro63-8695 GSK2315698
Solubility Information DMSO : 75 mg/mL (220.35 mM; Need ultrasonic)
SMILES O=C(N1[C@H](CCC1)C(O)=O)CCCCC(N2[C@H](CCC2)C(O)=O)=O
Molecular Weight (g/mol) 340.37
Formula Weight 340.37
Percent Purity 98.05%
Grade Research
Physical Form Solid
For Use With (Application) Neuroscience-Neuromodulation