Learn More
Miquelianin, MedChemExpress
Product Code. 16483396
Encompass_Preferred
Click to view available options
Quantity:
1 mg
5 mg
10 mg
25 mg
50 mg
Unit Size:
10mg
1mg
25mg
50mg
5mg
Product Code. 16483396
Brand: Medchem Express HY13930
Please to purchase this item. Need a web account? Register with us today!
MedChemExpress Miquelianin (Quercetin 3-O-glucuronide) is a metabolite of quercetin and a type of natural flavonoid.
Miquelianin (Quercetin 3-O-glucuronide) is a metabolite of quercetin and a type of natural flavonoid.
Specifications
Chemical Name or Material Miquelianin
CAS 22688-79-5
Color Yellow
Purity Grade Notes Research
Molecular Formula C21H18O13
Quantity 25 mg
Synonym Quercetin 3-O-glucuronide Quercetin 3-glucuronide
Solubility Information DMSO : ≥ 30 mg/mL (62.71 mM)
SMILES O=C1C2=C(O)C=C(O)C=C2OC(C3=CC=C(O)C(O)=C3)=C1O[C@@H]4O[C@H](C(O)=O)[C@@H](O)[C@H](O)[C@H]4O
Molecular Weight (g/mol) 478.36
Formula Weight 478.36
Percent Purity 98.3%
Grade Research
Physical Form Solid
For Use With (Application) Cancer-Kinase/protease