Learn More
MiPLA, TRC
Specifications
Specifications
| Chemical Name or Material | Methylisopropyllysergamide |
| CAS | 100768-08-9 |
| Molecular Formula | C20 H25 N3 O |
| Quantity | 1 mg |
| Synonym | (8β)-9,10-didehydro-N,6-dimethyl-N-(1-methylethyl)-ergoline-8-carboxamide, MiPLA (solution) |
| InChI Formula | InChI=1S/C20H25N3O/c1-12(2)23(4)20(24)14-8-16-15-6-5-7-17-19(15)13(10-21-17)9-18(16)22(3)11-14/h5-8,10,12,14,18,21H,9,11H2,1-4H3/t14-,18-/m1/s1 |
| SMILES | CC(C)N(C)C(=O)[C@H]1CN(C)[C@@H]2Cc3c[nH]c4cccc(C2=C1)c34 |
| IUPAC Name | (6aR,9R)-N-isopropyl-N,7-dimethyl-6,6a,8,9-tetrahydro-4H-indolo[4,3-fg]quinoline-9-carboxamide |
| Molecular Weight (g/mol) | 323.43 |
| Formula Weight | 323.1998 |