Learn More
Minoxidil-d10, TRC
Product Code. 30470807
Change view
Click to view available options
Quantity:
10 mg
Unit Size:
10mg
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Product Code. Quantity Unit Size
30470807 10 mg 10mg
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
Regulatory: Restrictions may apply to the purchase of this product and we may need to request additional documentation from you
Product Code. 30470807 Supplier Toronto Research Chemicals Supplier No. TRCM34500210MG
 Restricted Item
Estimated Shipment: 13-11-2026
Add to basket
Add to basket
missing translation for 'validMainframeMsg'
Regulatory: Restrictions may apply to the purchase of this product and we may need to request additional documentation from you
High-purity organic molecules and analytical standards, strategically delivered worldwide to empower innovation and commercial success.
TRUSTED_SUSTAINABILITY
Specifications
Chemical Name or Material Minoxidil-d10
CAS 1020718-66-4
Molecular Formula C9 2H10 H5 N5 O
Quantity 10 mg
Synonym 2,4-Pyrimidinediamine, 6-(1-piperidinyl-2,2,3,3,4,4,5,5,6,6-d10)-, 3-oxide (ACI), 6-(1-Piperidinyl-2,2,3,3,4,4,5,5,6,6-d10)-2,4-pyrimidinediamine 3-oxide, 6-(1-Piperidinyl-d10)-2,4-pyrimidinediamine 3-oxide, Minoxidil-d10, Minoxidil-D10 (piperidine-D10)
InChI Formula InChI=1S/C9H15N5O/c10-7-6-8(12-9(11)14(7)15)13-4-2-1-3-5-13/h6H,1-5,10H2,(H2,11,12)/i1D2,2D2,3D2,4D2,5D2
SMILES [2H]C1([2H])N(c2cc(N)[n+]([O-])c(N)n2)C([2H])([2H])C([2H])([2H])C([2H])([2H])C1([2H])[2H]
IUPAC Name 6-(2,2,3,3,4,4,5,5,6,6-decadeuteriopiperidin-1-yl)-3-oxidopyrimidin-3-ium-2,4-diamine
Molecular Weight (g/mol) 219.31
Formula Weight 219.1904