missing translation for 'onlineSavingsMsg'
Learn More

MINK1 Rabbit anti-Human, Mouse, Rat, Polyclonal, Proteintech

Product Code. 16861204 Shop All Proteintech Products
Change view
Click to view available options
Quantity:
20 μL
150 μL
Unit Size:
150µL
20µL
2 product options available for selection
Product selection table with 2 available options. Use arrow keys to navigate and Enter or Space to select.
Product Code. Quantity unitSize
16861204 20 μL 20µL
16851204 150 μL 150µL
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 2 options available.
2 options
This item is not returnable. View return policy
Product Code. 16861204 Supplier Proteintech Supplier No. 131371AP20UL

Please to purchase this item. Need a web account? Register with us today!

This item is not returnable. View return policy

Rabbit Polyclonal Antibody

This gene encodes a serine/threonine kinase belonging to the germinal center kinase (GCK) family. The protein is structurally similar to the kinases that are related to NIK and may belong to a distinct subfamily of NIK-related kinases within the GCK family. Studies of the mouse homolog indicate an up-regulation of expression in the course of postnatal mouse cerebral development and activation of the cJun N-terminal kinase (JNK) and the p38 pathways.

Specifications

Antigen MINK1
Applications Immunoprecipitation, Western Blot, Immunohistochemistry (Paraffin)
Classification Polyclonal
Concentration 0.14 mg/mL
Conjugate Unconjugated
Formulation PBS with 50% glycerol and 0.02% sodium azide; pH 7.3
Gene MINK1
Gene Accession No. F1LP90, Q8N4C8, Q9JM52
Gene Alias B55, GCK family kinase MiNK, hMINK, hMINKbeta, MAP4K6, MEK kinase kinase 6, MEKKK 6, MINK, MINK1, Misshapen like kinase 1, Misshapen/NIK related kinase, YSK2, ZC3
Gene Symbols Mink1
Host Species Rabbit
Immunogen MINK1 Fusion Protein Ag3795
Purification Method Antigen Affinity Chromatography
Quantity 20 μL
Regulatory Status RUO
Primary or Secondary Primary
Gene ID (Entrez) 303259, 50488, 50932
Target Species Rat, Mouse, Human
Content And Storage -20°C
Product Type Antibody
Form Liquid
Isotype IgG
Show More Show Less
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.