missing translation for 'onlineSavingsMsg'
Learn More

Thermo Scientific™ Minerals Modified Glutamate Medium (Dehydrated)

Product Code. 10473803
Change view
Click to view available options
Form:
Powder
Product Type:
Dehydrated Culture Media
Unit Size:
500g
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Product Code. Form Product Type unitSize
10473803 Powder Dehydrated Culture Media 500g
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
This item is not returnable. View return policy
Product Code. 10473803 Supplier Thermo Scientific™ Supplier No. CM0607G

Please to purchase this item. Need a web account? Register with us today!

This item is not returnable. View return policy

Identify members of the coliform group of bacteria in food and water samples with this medium.

Identify members of the coliform group of bacteria in food and water samples with Thermo Scientific™ Oxoid™ Minerals Modified Glutamate Medium (Dehydrated). The mineral content of the medium allows for the superior cultivation and enumeration of coliforms.

Use Minerals Modified Glutamate Medium to identify coliforms in food and water samples by the technique known as the Multiple Tube Method, Dilution Method, or the Most Probable Number (MPN) method.
  • Superior performance over MacConkey Broth in the detection of Escherichia coli.
  • Medium of choice for detection of small numbers of Escherichia coli in chlorinated water.
  • Effectively isolates Escherichia coli from sea water.

Minerals Modified Glutamate Medium was developed by Oxoid Laboratories and Metropolitan Water Board Laboratories and based upon the formulation described by Gray1

Not all products are available for sale in all territories. Please inquire.
Remel™ and Oxoid™ products are now part of the Thermo Scientific™ brand.
TRUSTED_SUSTAINABILITY

Specifications

Content And Storage 10°C to 30°C
Form Powder
Product Type Dehydrated Culture Media
Quantity Each
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.