Learn More
Merck Millipore Millipak™ Sterile Filters for Milli-Q™ IQ 7000 Ultrapure Water System
Purification cartridges
Brand: Merck Millipore MPGP002A1
This item is not returnable.
View return policy
Description
- 0.22μm membrane filter for particulate-free and bacteria-free water at the point of dispense for the Milli-Q IQ 7000 system
- Sterile unit
- Delivered hermetically sealed.