Learn More
Merck Millipore Millipak™ Sterile Filters for Milli-Q™ IQ 7000 Ultrapure Water System
Purification cartridges
Brand:  Merck Millipore MPGP002A1
 View more versions of this product
Product Code. 15749618
  • 589.85 € / Pack of 1
Please to purchase this item. Need a web account? Register with us today!
This item is not returnable. View return policy
Description
Description
  • 0.22μm membrane filter for particulate-free and bacteria-free water at the point of dispense for the Milli-Q IQ 7000 system
  • Sterile unit
  • Delivered hermetically sealed.