Learn More
Melford Miller's LB Broth, 25G Capsules
Description
RPI Media is now available in convenient pre-measured and pre-mixed capsules.
RPI Media is now available in convenient pre-measured and pre-mixed capsules. Prebuffered media eliminates messy weighing and pH adjustments.
Simply dissolve 1 capsule per 1 Liter of distilled water. Water must be 37°C or warmer to completely dissolve gelatin capsule. Agitate. Sterilize by autoclaving for 15 minutes before use. Buffer is set for 7.2 at 20°C using a 10mM solution of Tris/Tris HCl. pH will vary depending on temperature.
For maintenance and propagation of E. Coli. High Salt Formula. Molecular Biology Grade.
Ingredients:
Tryptone: 10 g/L
Sodium Chloride: 10 g/L
Yeast Extract: 5 g/L
Tris/Tris HCl: 1.5 g/L
Specifications
Specifications
| Quantity | 500 g |
Your input is important to us. Please complete this form to provide feedback related to the content on this product.