missing translation for 'onlineSavingsMsg'
Learn More

Milademetan, MedChemExpress

Product Code. p-200097060
Encompass_Preferred
Click to view available options
Quantity:
1 mg
5 mg
10 mg
Unit Size:
10mg
1mg
5mg
This item is not returnable. View return policy

Product Code. 16493308

Brand: Medchem Express HY101266

Please to purchase this item. Need a web account? Register with us today!

This item is not returnable. View return policy

MedChemExpress Milademetan (DS-3032) is a specific and orally active MDM2 inhibitor for the research of acute myeloid leukemia (AML) or solid tumors. Milademetan (DS-3032) induces G1 cell cycle arrest, senescence and apoptosis.

Milademetan (DS-3032) is a specific and orally active MDM2 inhibitor for the research of acute myeloid leukemia (AML) or solid tumors. Milademetan (DS-3032) induces G1 cell cycle arrest, senescence and apoptosis.

Specifications

Chemical Name or Material Milademetan
CAS 1398568-47-2
Color Off-White
Purity Grade Notes Research
Molecular Formula C30H34Cl2FN5O4
Quantity 10 mg
Synonym DS-3032
Solubility Information DMSO : 16.67 mg/mL (26.95 mM; Need ultrasonic)
SMILES O=C(NC1=CC(Cl)=CC=C21)[C@]32C4(CCC(C)(C)CC4)N[C@@H](C(N[C@@H]5CC[C@@H](C(N)=O)OC5)=O)[C@@H]3C6=CC=NC(Cl)=C6F
Molecular Weight (g/mol) 618.53
Formula Weight 618.53
Percent Purity 92.38%
Grade Research
Physical Form Powder
For Use With (Application) Cancer-programmed cell death
Show More Show Less

Research purposes only

Product Content Correction

The Fisher Scientific Encompass Program offers items which are not part of our distribution portfolio. These products typically do not have pictures or detailed descriptions. However, we are committed to improving your shopping experience. Please use the form below to provide feedback related to the content on this product.

Product Title

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.

Thank You! Your feedback has been submitted. Fisher Scientific is always working to improve our content for you. We appreciate your feedback.