missing translation for 'onlineSavingsMsg'
Learn More
Learn More
Invitrogen™ MICS1 Monoclonal Antibody (42), Invitrogen™

Mouse Monoclonal Antibody
Brand: Invitrogen™ MA543048
This item is not returnable.
View return policy
Description
Specificity: Dermatophagoides pteronyssinus GHITM/MICS1
Required for the mitochondrial tubular network and cristae organization. Involved in apoptotic release of cytochrome c.
Specifications
| MICS1 | |
| Monoclonal | |
| 1 mg/mL | |
| PBS with no preservative | |
| Mouse | |
| Protein A | |
| RUO | |
| Arthropod | |
| Antibody | |
| IgG2a |
| ELISA | |
| 42 | |
| Unconjugated | |
| 1010001P14Rik; C77840; dermal papilla-derived protein 2; DERP2; DKFZp566C0746; FLJ26584; GHITM; growth hormone inducible transmembrane protein; growth hormone-inducible transmembrane protein; growth hormone-inducible transmembrane protein-like protein; HSPC282; MICS1; mitochondrial morphology and cristae structure 1; My021; PTD010; similar to Dermal papilla derived protein 2; Tmbim5; transmembrane BAX inhibitor motif containing 5; transmembrane BAX inhibitor motif-containing protein 5; UNQ244/PRO281 | |
| Recombinant Dermatophagoides pteronyssinus GHITM/MICS1 Protein. | |
| 100 μL | |
| Primary | |
| -20°C, Avoid Freeze/Thaw Cycles | |
| Liquid |
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title
Spot an opportunity for improvement?Share a Content Correction