missing translation for 'onlineSavingsMsg'
Learn More
Learn More
Microsomal Glutathione S-transferase 1 Antibody, Novus Biologicals™
Rabbit Polyclonal Antibody
Brand: Novus Biologicals NBP2-97486-50ul
This item is not returnable.
View return policy
Description
Microsomal Glutathione S-transferase 1 Polyclonal antibody specifically detects Microsomal Glutathione S-transferase 1 in Human samples. It is validated for Immunohistochemistry (Paraffin)
Specifications
| Microsomal Glutathione S-transferase 1 | |
| Polyclonal | |
| Immunohistochemistry-Paraffin 1:50-1:200 | |
| glutathione S-transferase 12, GST12EC 2.5.1.18, MGC14525, MGST, MGST-I, microsomal glutathione S-transferase 1, Microsomal GST-1, Microsomal GST-I | |
| Produced in rabbits immunized with E. coli-derived Human Microsomal Glutathione S-transferase 1 fragment. | |
| 50 μg | |
| Cell Biology, Endocrinology | |
| 4257 | |
| Store at 4°C short term. Aliquot and store at -20°C long term. Avoid freeze-thaw cycles. | |
| IgG |
| Immunohistochemistry (Paraffin) | |
| Unconjugated | |
| PBS (pH 7.0) | |
| Rabbit | |
| Antigen and protein A Affinity-purified | |
| RUO | |
| Primary | |
| Human | |
| Purified |
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title
Spot an opportunity for improvement?Share a Content Correction