Learn More
behr Labor-Technik™ Microseparator
Product Code. 10156111
Click to view available options
:
Certification: IAW DIN 38407
This item is not returnable. View return policy
Product Code. 10156111
Brand: behr Labor-Technik™ B00217656
Only null left
Add to basket
Add to basket
This item is not returnable. View return policy
MICROSEPARATOR FOR DET'N OF VOLATILE HALOHYDROCARBONS, STD TAPER NS 19, IAW DIN 38407 F 4 F 4/19
Specifications
Description IAW DIN 38407