Learn More
Greiner Bio-One Microscopic Cover slip for Terasaki Plates
Product Code. 10206971
Click to view available options
:
Microscopic coverslip
This item is not returnable. View return policy
Product Code. 10206971
Brand: Greiner Bio-One 653081
Please to purchase this item. Need a web account? Register with us today!
This item is currently unavailable or has been discontinued.
View the product page for possible alternatives.
View alternative products
This item is not returnable. View return policy
For use with Terasaki plates. Greiner Bio-One Microscopic Cover slip is manufactured from crystal clear polystyrene.
  • Manufactured from crystal clear polystyrene
  • Non-sterile
TRUSTED_SUSTAINABILITY
Specifications
Material Polystyrene
Length (Metric) 76 mm
Width (Metric) 51 mm
For Use With (Equipment) Terasaki Plates